Product Details
- SNP ID
-
rs12140499
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
40
- Location
-
Chr.1:219177506 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAAGACCTTGTATGCATTACAAAGA[A/G]GCACCTCACACTCAACCTGTGTATG
- Phenotype
-
MIM: 616548
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LYPLAL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111275274] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LYPLAL1
- Gene Name
- lysophospholipase like 1
- Gene
- LYPLAL1-AS1
- Gene Name
- LYPLAL1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details