Product Details

SNP ID
rs7056350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:81205570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATTCACCGTTGATGGACACTTA[C/G]CCACATATCATAGTTATTTTGAATA
Phenotype
MIM: 300385 MIM: 300190
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HMGN5 PubMed Links

Gene Details

Gene
HMGN5
Gene Name
high mobility group nucleosome binding domain 5
There are no transcripts associated with this gene.

Gene
SH3BGRL
Gene Name
SH3 domain binding glutamate rich protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003022.2 Intron NP_003013.1
XM_011531013.1 Intron XP_011529315.1
XM_011531014.1 Intron XP_011529316.1

View Full Product Details