Product Details

SNP ID
rs11101821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133554151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATTTAAAAGTCATGTTTGATAGT[A/G]TATTGCCAGTTTGTTTAAGACAGGT
Phenotype
MIM: 611486
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYCE1 PubMed Links

Gene Details

Gene
SYCE1
Gene Name
synaptonemal complex central element protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143763.1 1201 UTR 3 NP_001137235.1
NM_001143764.1 1201 Intron NP_001137236.1
NM_130784.2 1201 UTR 3 NP_570140.1

View Full Product Details