Product Details

SNP ID
rs4420479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:66514179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTACACGTTTTAAAATCAGCATC[A/G]GTGAAAATGAGTTCTCATTCTTTTA
Phenotype
MIM: 180479 MIM: 609984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL4 PubMed Links

Gene Details

Gene
RPL4
Gene Name
ribosomal protein L4
There are no transcripts associated with this gene.

Gene
ZWILCH
Gene Name
zwilch kinetochore protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287821.1 Intron NP_001274750.1
NM_001287822.1 Intron NP_001274751.1
NM_001287823.1 Intron NP_001274752.1
NM_017975.4 Intron NP_060445.3

View Full Product Details