Product Details

SNP ID
rs12290656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:103909141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGCCTTTTCTCTGGAAACCATA[C/T]TTTTCCTTTTTTAATAATCAACTAA
Phenotype
MIM: 609673
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDGFD PubMed Links

Gene Details

Gene
PDGFD
Gene Name
platelet derived growth factor D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025208.4 2020 UTR 3 NP_079484.1
NM_033135.3 2020 UTR 3 NP_149126.1

View Full Product Details