Product Details

SNP ID
rs8109430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11342647 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAAATGGGCATCATTATAGGAGA[A/G]GCTTACAGGATAAAATGGCTGACAC
Phenotype
MIM: 604350 MIM: 613771
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC159 PubMed Links

Gene Details

Gene
CCDC159
Gene Name
coiled-coil domain containing 159
There are no transcripts associated with this gene.

Gene
RAB3D
Gene Name
RAB3D, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
TMEM205
Gene Name
transmembrane protein 205
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145416.1 736 Intron NP_001138888.1
NM_001321112.1 736 Intron NP_001308041.1
NM_001321113.1 736 Intron NP_001308042.1
NM_001321114.1 736 Intron NP_001308043.1
NM_033408.3 736 Intron NP_212133.1
NM_198536.2 736 Intron NP_940938.1
XM_005259899.2 736 Intron XP_005259956.1
XM_017026769.1 736 Missense Mutation CTC,TTC L240F XP_016882258.1

View Full Product Details