Product Details

SNP ID
rs11011636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:19821406 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTCTCCACAAAAGGACGTGTTGT[C/T]TTAGAATTCTCAGTGCACTTTATGC
Phenotype
MIM: 606827
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLXDC2 PubMed Links

Gene Details

Gene
PLXDC2
Gene Name
plexin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282736.1 Intron NP_001269665.1
NM_032812.8 Intron NP_116201.7
XM_011519750.2 Intron XP_011518052.1

View Full Product Details