Product Details

SNP ID
rs13193160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49512108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATTTTATTTTATTTCTTTATTG[G/T]GTTAATCTGTAAAATAATGTCAAAC
Phenotype
MIM: 614763
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GLYATL3 PubMed Links

Gene Details

Gene
GLYATL3
Gene Name
glycine-N-acyltransferase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010904.1 Intron NP_001010904.1

View Full Product Details