Product Details

SNP ID
rs12626491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:32779971 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAGAGTACCCCGTGAGTTAGGA[A/G]GCCAGGTGATATGGTTTGAATATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C21orf62-AS1 PubMed Links

Gene Details

Gene
C21orf62-AS1
Gene Name
C21orf62 antisense RNA 1
There are no transcripts associated with this gene.

Gene
PAXBP1
Gene Name
PAX3 and PAX7 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details