Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001109977.1 | 9999 | Intron | NP_001103447.1 | ||
XM_005263199.1 | 9999 | UTR 3 | XP_005263256.1 | ||
XM_011532220.1 | 9999 | UTR 3 | XP_011530522.1 | ||
XM_011532221.2 | 9999 | UTR 3 | XP_011530523.1 | ||
XM_011532222.2 | 9999 | UTR 3 | XP_011530524.1 | ||
XM_011532223.2 | 9999 | UTR 3 | XP_011530525.1 | ||
XM_011532224.2 | 9999 | UTR 3 | XP_011530526.1 | ||
XM_017008583.1 | 9999 | UTR 3 | XP_016864072.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004564.2 | 9999 | Intron | NP_004555.1 | ||
XM_005263063.3 | 9999 | Intron | XP_005263120.1 |