Product Details

SNP ID
rs4992171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40182293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTGCTTTGGAGGCTGCAGAGCC[A/C]TGTACAAATGAAAGGTGTTATTTAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RAPGEFL1 PubMed Links

Gene Details

Gene
RAPGEFL1
Gene Name
Rap guanine nucleotide exchange factor like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303533.1 Intron NP_001290462.1
NM_001303534.1 Intron NP_001290463.1
NM_016339.4 Intron NP_057423.1
XM_006721932.3 Intron XP_006721995.1
XM_006721935.3 Intron XP_006721998.1
XM_006721937.3 Intron XP_006722000.1
XM_006721938.3 Intron XP_006722001.1

View Full Product Details