Product Details

SNP ID
rs16852892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:143010376 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTACCTTTTAATAGACAGCAAG[A/G]ATTTATATGTGGATTTCTTTATGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100289361 PubMed Links

Gene Details

Gene
LOC100289361
Gene Name
uncharacterized LOC100289361
There are no transcripts associated with this gene.

Gene
PAQR9-AS1
Gene Name
PAQR9 antisense RNA 1
There are no transcripts associated with this gene.

Gene
U2SURP
Gene Name
U2 snRNP associated SURP domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080415.1 Intron NP_001073884.1
NM_001320219.1 Intron NP_001307148.1
NM_001320220.1 Intron NP_001307149.1
NM_001320222.1 Intron NP_001307151.1
XM_005247248.4 Intron XP_005247305.1
XM_017006037.1 Intron XP_016861526.1
XM_017006038.1 Intron XP_016861527.1
XM_017006039.1 Intron XP_016861528.1

View Full Product Details