Product Details

SNP ID
rs16957784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68254071 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGCAGGAGCTGGATGCAAGGAG[A/T]TGGAGCGATGGAAAGCTGAAAGAGG
Phenotype
MIM: 609362 MIM: 605641
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G15 PubMed Links

Gene Details

Gene
PLA2G15
Gene Name
phospholipase A2 group XV
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012320.3 Intron NP_036452.1
XM_005255886.3 Intron XP_005255943.1
XM_011522979.2 Intron XP_011521281.1
XM_011522980.2 Intron XP_011521282.2
Gene
SLC7A6
Gene Name
solute carrier family 7 member 6
There are no transcripts associated with this gene.

View Full Product Details