Product Details
- SNP ID
-
rs35565861
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:179443448 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGGGCCAAAGGCTTCATCATATC[A/T]TTCTGGAATGCAAGTTTTGCCTAAG
- Phenotype
-
MIM: 612344
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF385B
PubMed Links
Gene Details
- Gene
- ZNF385B
- Gene Name
- zinc finger protein 385B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001113397.1 |
1138 |
Missense Mutation |
AAA,AAT |
K330N |
NP_001106868.1 |
NM_001113398.1 |
1138 |
Missense Mutation |
AAA,AAT |
K304N |
NP_001106869.1 |
NM_001282725.1 |
1138 |
Missense Mutation |
AAA,AAT |
K304N |
NP_001269654.1 |
NM_152520.4 |
1138 |
Missense Mutation |
AAA,AAT |
K406N |
NP_689733.3 |
XM_011510713.2 |
1138 |
Missense Mutation |
AAA,AAT |
K461N |
XP_011509015.1 |
XM_011510714.2 |
1138 |
Missense Mutation |
AAA,AAT |
K448N |
XP_011509016.1 |
XM_011510715.2 |
1138 |
Missense Mutation |
AAA,AAT |
K434N |
XP_011509017.1 |
XM_011510716.2 |
1138 |
Missense Mutation |
AAA,AAT |
K364N |
XP_011509018.1 |
XM_011510717.2 |
1138 |
Missense Mutation |
AAA,AAT |
K364N |
XP_011509019.1 |
XM_011510719.2 |
1138 |
Missense Mutation |
AAA,AAT |
K344N |
XP_011509021.1 |
XM_011510720.2 |
1138 |
Missense Mutation |
AAA,AAT |
K344N |
XP_011509022.1 |
XM_011510721.2 |
1138 |
Missense Mutation |
AAA,AAT |
K331N |
XP_011509023.1 |
XM_011510723.2 |
1138 |
Intron |
|
|
XP_011509025.1 |
XM_017003435.1 |
1138 |
Missense Mutation |
AAA,AAT |
K421N |
XP_016858924.1 |
XM_017003436.1 |
1138 |
Missense Mutation |
AAA,AAT |
K344N |
XP_016858925.1 |
XM_017003437.1 |
1138 |
Missense Mutation |
AAA,AAT |
K338N |
XP_016858926.1 |
XM_017003438.1 |
1138 |
Missense Mutation |
AAA,AAT |
K304N |
XP_016858927.1 |
XM_017003439.1 |
1138 |
Missense Mutation |
AAA,AAT |
K304N |
XP_016858928.1 |
XM_017003440.1 |
1138 |
Missense Mutation |
AAA,AAT |
K292N |
XP_016858929.1 |
XM_017003441.1 |
1138 |
Intron |
|
|
XP_016858930.1 |
View Full Product Details