Product Details

SNP ID
rs17677608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:4081028 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACATTGTAATTATTCATTTACAAC[A/G]TCTCAATCACTTTCTTTTCAACACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf201 PubMed Links

Gene Details

Gene
C6orf201
Gene Name
chromosome 6 open reading frame 201
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085401.2 Intron NP_001078870.1
XM_005249112.3 Intron XP_005249169.1
XM_011514613.2 Intron XP_011512915.1
XM_017010861.1 Intron XP_016866350.1
Gene
FAM217A
Gene Name
family with sequence similarity 217 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173563.2 Intron NP_775834.2
XM_005248951.3 Intron XP_005249008.1
XM_006715025.2 Intron XP_006715088.1
XM_006715026.3 Intron XP_006715089.1
XM_011514413.2 Intron XP_011512715.1
XM_011514414.2 Intron XP_011512716.1
XM_011514415.1 Intron XP_011512717.1
XM_011514416.2 Intron XP_011512718.1
XM_011514417.1 Intron XP_011512719.1
XM_017010480.1 Intron XP_016865969.1
XM_017010481.1 Intron XP_016865970.1
XM_017010482.1 Intron XP_016865971.1

View Full Product Details