Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000564.4 | 2311 | UTR 3 | NP_000555.2 | ||
NM_001243099.1 | 2311 | UTR 3 | NP_001230028.1 | ||
NM_175724.2 | 2311 | Intron | NP_783851.1 | ||
NM_175725.2 | 2311 | Intron | NP_783852.1 | ||
NM_175726.3 | 2311 | UTR 3 | NP_783853.1 | ||
NM_175727.2 | 2311 | Intron | NP_783854.1 | ||
NM_175728.2 | 2311 | Intron | NP_783855.1 | ||
XM_011533677.2 | 2311 | UTR 3 | XP_011531979.1 | ||
XM_011533678.2 | 2311 | UTR 3 | XP_011531980.1 |