Product Details

SNP ID
rs16950553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67069447 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAAGCATTTGCCGCTGAATGAGA[A/G]TTGGTGTGTGAGCGTTATATGAGGA
Phenotype
MIM: 603109
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102723493 PubMed Links

Gene Details

Gene
LOC102723493
Gene Name
uncharacterized LOC102723493
There are no transcripts associated with this gene.

Gene
SMAD3
Gene Name
SMAD family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145102.1 Intron NP_001138574.1
NM_001145103.1 Intron NP_001138575.1
NM_001145104.1 Intron NP_001138576.1
NM_005902.3 Intron NP_005893.1
XM_011521559.2 Intron XP_011519861.1
XM_011521560.1 Intron XP_011519862.1

View Full Product Details