Product Details

SNP ID
rs16973717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37559617 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATCTGGACTCAAACCAACTATTA[C/T]TGTGAAAGGTTGACTTATCTGTTCA
Phenotype
MIM: 613903
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF540 PubMed Links

Gene Details

Gene
ZNF540
Gene Name
zinc finger protein 540
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172225.2 Intron NP_001165696.1
NM_001172226.2 Intron NP_001165697.1
NM_152606.4 Intron NP_689819.1
Gene
ZNF571
Gene Name
zinc finger protein 571
There are no transcripts associated with this gene.

Gene
ZNF571-AS1
Gene Name
ZNF571 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details