Product Details

SNP ID
rs35602244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100352429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGAGCTCATCGTCTGTGGAGCC[A/C]CCGCTCTTACAAAGCTCGGCTGCCC
Phenotype
MIM: 604532 MIM: 604031
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PKD2L1 PubMed Links

Gene Details

Gene
PKD2L1
Gene Name
polycystin 2 like 1, transient receptor potential cation channel
There are no transcripts associated with this gene.

Gene
SCD
Gene Name
stearoyl-CoA desaturase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005063.4 864 Missense Mutation CAC,CCC H125P NP_005054.3

View Full Product Details