Product Details

SNP ID
rs17754462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27547322 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAGATTTTTAATGCTTTTGGGCT[C/G]TTCAAGTAAGTGCAGCTTTACCATC
Phenotype
MIM: 609733
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LNX2 PubMed Links

Gene Details

Gene
LNX2
Gene Name
ligand of numb-protein X 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153371.3 3254 UTR 3 NP_699202.1
XM_011534995.2 3254 UTR 3 XP_011533297.1
XM_017020434.1 3254 UTR 3 XP_016875923.1

View Full Product Details