Product Details

SNP ID
rs16892479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:161347581 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGTGCAAATAATACTGTGATTCA[C/T]GTTCATGTGTAGTGGATGATCTTGC
Phenotype
MIM: 602544
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARK2 PubMed Links

Gene Details

Gene
PARK2
Gene Name
parkin RBR E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004562.2 4047 UTR 3 NP_004553.2
NM_013987.2 4047 UTR 3 NP_054642.2
NM_013988.2 4047 UTR 3 NP_054643.2
XM_011535863.1 4047 UTR 3 XP_011534165.1
XM_017010908.1 4047 UTR 3 XP_016866397.1
XM_017010909.1 4047 UTR 3 XP_016866398.1

View Full Product Details