Product Details

SNP ID
rs17081549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:151654501 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTACTGGTTCTCATCTGTCACATA[G/T]GGAAAAATTATAGAAGGAAAACATC
Phenotype
MIM: 133430
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ESR1 PubMed Links

Gene Details

Gene
ESR1
Gene Name
estrogen receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000125.3 354 Intron NP_000116.2
NM_001122740.1 354 Intron NP_001116212.1
NM_001122741.1 354 Intron NP_001116213.1
NM_001122742.1 354 Intron NP_001116214.1
NM_001291230.1 354 Intron NP_001278159.1
NM_001291241.1 354 Intron NP_001278170.1
NM_001328100.1 354 Intron NP_001315029.1
XM_006715374.3 354 Intron XP_006715437.1
XM_006715375.3 354 Intron XP_006715438.1
XM_011535543.2 354 Intron XP_011533845.1
XM_011535544.2 354 Intron XP_011533846.1
XM_011535545.2 354 Intron XP_011533847.1
XM_011535547.2 354 Intron XP_011533849.1
XM_011535549.2 354 Intron XP_011533851.1
XM_017010376.1 354 UTR 5 XP_016865865.1
XM_017010377.1 354 UTR 5 XP_016865866.1
XM_017010378.1 354 Intron XP_016865867.1
XM_017010379.1 354 Intron XP_016865868.1
XM_017010380.1 354 UTR 5 XP_016865869.1
XM_017010381.1 354 Intron XP_016865870.1
XM_017010382.1 354 Intron XP_016865871.1
XM_017010383.1 354 Intron XP_016865872.1

View Full Product Details