Product Details
- SNP ID
-
rs8032961
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:77430817 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAGCCAATAGCTTTGGATAACTT[C/G]TGACAGCAACAATATGAAACTATTA
- Phenotype
-
MIM: 605534
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HMG20A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115924885] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HMG20A
- Gene Name
- high mobility group 20A
View Full Product Details