Product Details

SNP ID
rs12974003
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38412086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGTTTGGAGATTATCCAGGTTT[G/T]TTGTTGTTGTTGTTGTTGTTGTTGT
Phenotype
MIM: 607320
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM98C PubMed Links

Gene Details

Gene
FAM98C
Gene Name
family with sequence similarity 98 member C
There are no transcripts associated with this gene.

Gene
RASGRP4
Gene Name
RAS guanyl releasing protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146202.1 Intron NP_001139674.1
NM_001146203.1 Intron NP_001139675.1
NM_001146204.1 Intron NP_001139676.1
NM_001146205.1 Intron NP_001139677.1
NM_001146206.1 Intron NP_001139678.1
NM_001146207.1 Intron NP_001139679.1
NM_170604.2 Intron NP_733749.1

View Full Product Details