Product Details
- SNP ID
-
rs7588239
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:88073825 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGTATTGTTTTAACTTGAATTTTT[G/T]ATTATTAGAGAAGTGAAAGTTTTTT
- Phenotype
-
MIM: 606846
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4780
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140796387] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR4780
- Gene Name
- microRNA 4780
There are no transcripts associated with this gene.
- Gene
- SMYD1
- Gene Name
- SET and MYND domain containing 1
View Full Product Details