Product Details

SNP ID
rs11541813
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:58681223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTACTGGAGCCTGAAGGAAGCC[C/T]CAGCCTGTGTCTGCTGAAGTTAATG
Phenotype
MIM: 604860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927322 PubMed Links

Gene Details

Gene
LOC101927322
Gene Name
uncharacterized LOC101927322
There are no transcripts associated with this gene.

Gene
MALT1
Gene Name
MALT1 paracaspase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006785.3 521 Missense Mutation CCC,CTC P88L NP_006776.1
NM_173844.2 521 Missense Mutation CCC,CTC P88L NP_776216.1
XM_011525794.1 521 Missense Mutation CCC,CTC P88L XP_011524096.1

View Full Product Details