Product Details

SNP ID
rs28901604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233924046 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAACTACTTTTACAGTGTTGGGG[A/G]GAAAGTGTTGAATGCAATAAGAAGA
Phenotype
MIM: 606678
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRPM8 PubMed Links

Gene Details

Gene
TRPM8
Gene Name
transient receptor potential cation channel subfamily M member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024080.4 Intron NP_076985.4
XM_011511810.2 Intron XP_011510112.1
XM_017004891.1 Intron XP_016860380.1

View Full Product Details