Product Details

SNP ID
rs28633784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:169821333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACTGGAAACATTCAGAATAGGCA[A/T]TTTTTTTTAACTGAAATTAGGCTGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LRRC34 PubMed Links

Gene Details

Gene
LRRC34
Gene Name
leucine rich repeat containing 34
There are no transcripts associated with this gene.

Gene
LRRIQ4
Gene Name
leucine rich repeats and IQ motif containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080460.1 Intron NP_001073929.1
XM_005247421.2 Intron XP_005247478.1
XM_006713613.3 Intron XP_006713676.1
XM_011512748.1 Intron XP_011511050.1
XM_011512749.1 Intron XP_011511051.1

View Full Product Details