Product Details

SNP ID
rs17874783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90213007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTATAATTTTTTTACTCCGATT[C/G]TCTCTTGGTTGCTTCAAAGCTTAGA
Phenotype
MIM: 605094
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STEAP2 PubMed Links

Gene Details

Gene
STEAP2
Gene Name
STEAP2 metalloreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040665.1 539 Intron NP_001035755.1
NM_001040666.1 539 Intron NP_001035756.1
NM_001244944.1 539 Intron NP_001231873.1
NM_001244945.1 539 Intron NP_001231874.1
NM_001244946.1 539 Intron NP_001231875.1
NM_152999.3 539 Intron NP_694544.2
XM_006715921.3 539 Intron XP_006715984.1
XM_017011953.1 539 Intron XP_016867442.1
XM_017011954.1 539 Intron XP_016867443.1
XM_017011955.1 539 Intron XP_016867444.1
XM_017011956.1 539 Intron XP_016867445.1
XM_017011957.1 539 Intron XP_016867446.1
XM_017011958.1 539 UTR 5 XP_016867447.1
Gene
STEAP2-AS1
Gene Name
STEAP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details