Product Details

SNP ID
rs28443180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99621006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAACTATTTCTCACTGGCCTGTTC[A/G]CTGCAGGGTCTAGGCTTTGCATTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN25 PubMed Links

Gene Details

Gene
ZSCAN25
Gene Name
zinc finger and SCAN domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145115.2 Intron NP_660090.2
XM_005250194.2 Intron XP_005250251.1
XM_011515905.2 Intron XP_011514207.1
XM_011515906.2 Intron XP_011514208.1
XM_011515907.2 Intron XP_011514209.1
XM_011515908.2 Intron XP_011514210.1
XM_011515909.2 Intron XP_011514211.1
XM_011515910.2 Intron XP_011514212.1
XM_017011824.1 Intron XP_016867313.1
XM_017011825.1 Intron XP_016867314.1
XM_017011826.1 Intron XP_016867315.1
XM_017011827.1 Intron XP_016867316.1

View Full Product Details