Product Details

SNP ID
rs34106027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113409043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAAGTGAGCCAGACGGACTGCA[A/G]GAGCTAATTACATTGTACAATTGGA
Phenotype
MIM: 125270 MIM: 607267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALAD PubMed Links

Gene Details

Gene
ALAD
Gene Name
aminolevulinate dehydratase
There are no transcripts associated with this gene.

Gene
C9orf43
Gene Name
chromosome 9 open reading frame 43
There are no transcripts associated with this gene.

Gene
POLE3
Gene Name
DNA polymerase epsilon 3, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278255.1 Intron NP_001265184.1
NM_017443.4 Intron NP_059139.3

View Full Product Details