Product Details

SNP ID
rs35072051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113410215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCTGCCCGTTCGTCCGCCCCCC[C/A]CGAGCTGCTCACCGCCTCCTTGATG
Phenotype
MIM: 125270 MIM: 607267
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
ALAD PubMed Links

Gene Details

Gene
ALAD
Gene Name
aminolevulinate dehydratase
There are no transcripts associated with this gene.

Gene
C9orf43
Gene Name
chromosome 9 open reading frame 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278629.1 Intron NP_001265558.1
NM_001278630.1 Intron NP_001265559.1
NM_152786.2 Intron NP_689999.1
XM_005251885.3 Intron XP_005251942.1
XM_011518477.1 Intron XP_011516779.1
XM_011518478.1 Intron XP_011516780.1
XM_011518479.2 Intron XP_011516781.1
Gene
POLE3
Gene Name
DNA polymerase epsilon 3, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278255.1 Intron NP_001265184.1
NM_017443.4 Intron NP_059139.3

View Full Product Details