Product Details

SNP ID
hCV60119913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65196253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATAAATAAGTTATTTACATTTC[A/C]AGTAAAAGTATTGTGTTTTATTTCT
Phenotype
MIM: 603489
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CILP PubMed Links

Gene Details

Gene
CILP
Gene Name
cartilage intermediate layer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003613.3 4214 UTR 3 NP_003604.3

View Full Product Details