Product Details

SNP ID
rs28367283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:14073244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAAAGACTCTCGTACCAGGCGA[A/G]TCTTCATTATACAGATCTGTCCCAC
Phenotype
MIM: 609463
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MKL2 PubMed Links

Gene Details

Gene
MKL2
Gene Name
MKL1/myocardin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308142.1 Intron NP_001295071.1
NM_014048.4 Intron NP_054767.3
XM_005255452.3 Intron XP_005255509.1
XM_005255453.4 Intron XP_005255510.1
XM_005255455.3 Intron XP_005255512.1
XM_006720908.3 Intron XP_006720971.1
XM_006720909.3 Intron XP_006720972.1
XM_006720912.3 Intron XP_006720975.1
XM_006720913.2 Intron XP_006720976.1
XM_006720914.2 Intron XP_006720977.1
XM_011522568.2 Intron XP_011520870.1
XM_011522569.2 Intron XP_011520871.1
XM_017023501.1 Intron XP_016878990.1
XM_017023502.1 Intron XP_016878991.1
XM_017023503.1 Intron XP_016878992.1
XM_017023504.1 Intron XP_016878993.1
XM_017023505.1 Intron XP_016878994.1

View Full Product Details