Product Details

SNP ID
rs28624377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:23652088 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCTAACAACTTTAGTTCTGGAT[A/T]ATTTTTTATACTCAGCATTCTGATT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF675 PubMed Links

Gene Details

Gene
ZNF675
Gene Name
zinc finger protein 675
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138330.2 3645 Intron NP_612203.2
XM_017026442.1 3645 UTR 3 XP_016881931.1
XM_017026443.1 3645 UTR 3 XP_016881932.1
XM_017026444.1 3645 Intron XP_016881933.1

View Full Product Details