Product Details

SNP ID
rs28738711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:118727654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAGAGAATGCCCTAGAAAAAC[A/G]TGTAACCAAATCTTAAGAGAATGTG
Phenotype
MIM: 607186
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC24D PubMed Links

Gene Details

Gene
SEC24D
Gene Name
SEC24 homolog D, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318066.1 Intron NP_001304995.1
NM_014822.3 Intron NP_055637.2
XM_005263379.2 Intron XP_005263436.1
XM_017008875.1 Intron XP_016864364.1

View Full Product Details