Product Details
- SNP ID
-
rs28631502
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:76992777 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTGCCCTCAAAAGTCGCGCATCC[C/G]GGGAATCCCCGTCTCCTGCAGTCCT
- Phenotype
-
MIM: 608102
MIM: 605653
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CLN5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139193170] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLN5
- Gene Name
- ceroid-lipofuscinosis, neuronal 5
- Gene
- FBXL3
- Gene Name
- F-box and leucine rich repeat protein 3
View Full Product Details