Product Details

SNP ID
rs28687504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:34433172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCTCAAGATTTCCTGTAAGAC[C/T]AAACTTTATCTTCCATATGTCTCAC
Phenotype
MIM: 613540
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPTSSA PubMed Links

Gene Details

Gene
SPTSSA
Gene Name
serine palmitoyltransferase small subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138288.3 2300 UTR 3 NP_612145.2

View Full Product Details