Product Details

SNP ID
rs35566516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:37598914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCATGCTTGGTGTGGAGTGGTTC[C/G]AGTTCCTCCGCGGGCCCCCACCCCC
Phenotype
MIM: 602294
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FOXA1 PubMed Links

Gene Details

Gene
FOXA1
Gene Name
forkhead box A1
There are no transcripts associated with this gene.

Gene
TTC6
Gene Name
tetratricopeptide repeat domain 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310135.1 Intron NP_001297064.1
XM_011537430.2 Intron XP_011535732.1
XM_011537431.2 Intron XP_011535733.1
XM_011537432.2 Intron XP_011535734.1
XM_017021254.1 Intron XP_016876743.1
XM_017021255.1 Intron XP_016876744.1
XM_017021256.1 Intron XP_016876745.1
XM_017021257.1 Intron XP_016876746.1

View Full Product Details