Product Details

SNP ID
rs17880632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64941049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATGAGCCCAGAGCTGATCTCTA[A/G]TGGACTTCTCTGTTTTTGTTTCTGT
Phenotype
MIM: 608577 MIM: 138319
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHURC1 PubMed Links

Gene Details

Gene
CHURC1
Gene Name
churchill domain containing 1
There are no transcripts associated with this gene.

Gene
CHURC1-FNTB
Gene Name
CHURC1-FNTB readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202558.1 Intron NP_001189487.1
NM_001202559.1 Intron NP_001189488.1
Gene
GPX2
Gene Name
glutathione peroxidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002083.3 Intron NP_002074.2
Gene
RAB15
Gene Name
RAB15, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details