Product Details

SNP ID
rs34894270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96114792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAACCCCACAGGGGCAGCGCAGG[C/T]GGGCTCACCAGGCCGTCTGGGTCCA
Phenotype
MIM: 104260 MIM: 608860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADRA2B PubMed Links

Gene Details

Gene
ADRA2B
Gene Name
adrenoceptor alpha 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000682.6 1454 UTR 3 NP_000673.2
Gene
ASTL
Gene Name
astacin-like metallo-endopeptidase (M12 family)
There are no transcripts associated with this gene.

View Full Product Details