Product Details

SNP ID
rs28585598
Assay Type
Functionally Tested
NCBI dbSNP Submissions
24
Location
Chr.1:37960682 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTTTTTTTTGAGACGGACTCTC[A/G]CTCTGTCGCCCAGGCTGGAGTGCAG
Phenotype
MIM: 605596
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SF3A3 PubMed Links
Additional Information
For this assay, SNP(s) [rs113878822] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
SF3A3
Gene Name
splicing factor 3a subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320830.1 Intron NP_001307759.1
NM_006802.3 Intron NP_006793.1

View Full Product Details