Product Details
- SNP ID
-
rs28429145
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:129756930 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGAGTGTCTAGTAAGGCTTTCC[A/C]CAGTAAAAAAAAAAAAAAAAAAAAA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1024L
PubMed Links
Gene Details
- Gene
- KIAA1024L
- Gene Name
- KIAA1024 like
View Full Product Details