Product Details

SNP ID
rs45614137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222298184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCGGGACAATTTCGAGACAACTT[C/G]GAGACAATTTCGAATGGACAAATTG
Phenotype
MIM: 606597
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC140 PubMed Links

Gene Details

Gene
CCDC140
Gene Name
coiled-coil domain containing 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153038.1 38 UTR 5 NP_694583.1
XM_017003466.1 38 Intron XP_016858955.1
Gene
PAX3
Gene Name
paired box 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000438.5 38 Intron NP_000429.2
NM_001127366.2 38 Intron NP_001120838.1
NM_013942.4 38 Intron NP_039230.1
NM_181457.3 38 Intron NP_852122.1
NM_181458.3 38 Intron NP_852123.1
NM_181459.3 38 Intron NP_852124.1
NM_181460.3 38 Intron NP_852125.1
NM_181461.3 38 Intron NP_852126.1

View Full Product Details