Product Details

SNP ID
rs41274005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33026343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAATTATCGCAGGAATTGTTAC[C/T]TTTTTGGTGAAATTTAGGGAAGGTG
Phenotype
MIM: 606350 MIM: 602837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APTX PubMed Links

Gene Details

Gene
APTX
Gene Name
aprataxin
There are no transcripts associated with this gene.

Gene
DNAJA1
Gene Name
DnaJ heat shock protein family (Hsp40) member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314039.1 Intron NP_001300968.1
NM_001539.3 Intron NP_001530.1

View Full Product Details