Product Details

SNP ID
rs41322845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43912903 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCAAAAATGGGAAAATCAGGCC[A/G]TATGACTAACTTATATATCCTCCAA
Phenotype
MIM: 194554
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF45 PubMed Links

Gene Details

Gene
ZNF45
Gene Name
zinc finger protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003425.3 3687 UTR 3 NP_003416.1
XM_011527267.1 3687 UTR 3 XP_011525569.1
XM_011527269.1 3687 UTR 3 XP_011525571.1
XM_011527271.1 3687 UTR 3 XP_011525573.1
XM_011527273.1 3687 UTR 3 XP_011525575.1
XM_017027217.1 3687 UTR 3 XP_016882706.1
XM_017027218.1 3687 UTR 3 XP_016882707.1
XM_017027219.1 3687 UTR 3 XP_016882708.1
XM_017027220.1 3687 UTR 3 XP_016882709.1
XM_017027221.1 3687 UTR 3 XP_016882710.1
XM_017027222.1 3687 UTR 3 XP_016882711.1
XM_017027223.1 3687 UTR 3 XP_016882712.1
XM_017027224.1 3687 UTR 3 XP_016882713.1
XM_017027225.1 3687 UTR 3 XP_016882714.1
XM_017027226.1 3687 UTR 3 XP_016882715.1
XM_017027227.1 3687 UTR 3 XP_016882716.1
XM_017027228.1 3687 UTR 3 XP_016882717.1
XM_017027229.1 3687 UTR 3 XP_016882718.1
XM_017027230.1 3687 UTR 3 XP_016882719.1

View Full Product Details