Product Details

SNP ID
rs41308124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23103060 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAACTCTGGGGACAGAGCGCCCC[G/T]GCCGCCTGATGGCCGAGGCAGGGTG
Phenotype
MIM: 603613
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC254896 PubMed Links

Gene Details

Gene
LOC254896
Gene Name
uncharacterized LOC254896
There are no transcripts associated with this gene.

Gene
TNFRSF10C
Gene Name
TNF receptor superfamily member 10c
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003841.3 247 UTR 5 NP_003832.2

View Full Product Details