Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000943.4 | Intron | NP_000934.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014035.2 | Intron | NP_054754.1 | |||
XM_005271972.4 | Intron | XP_005272029.1 | |||
XM_006714592.3 | Intron | XP_006714655.1 | |||
XM_011543349.2 | Intron | XP_011541651.1 | |||
XM_011543350.2 | Intron | XP_011541652.1 | |||
XM_011543352.1 | Intron | XP_011541654.1 | |||
XM_017009394.1 | Intron | XP_016864883.1 | |||
XM_017009395.1 | Intron | XP_016864884.1 | |||
XM_017009396.1 | Intron | XP_016864885.1 |