Product Details
- SNP ID
-
rs57188315
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:156669745 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGACTCCGTTTCCAGGGCAGTCTT[A/G]GGGGCACCAGCCACTGCACCCCTCA
- Phenotype
-
MIM: 600915
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NES
PubMed Links
Gene Details
- Gene
- NES
- Gene Name
- nestin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006617.1 |
4576 |
Silent Mutation |
CCC,CCT |
P1481P |
NP_006608.1 |
View Full Product Details