Product Details
- SNP ID
-
rs60130759
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:2270098 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGGGCAGACACCAGGAGCACCCT[A/G]CCTGCCTAGAGGACCACTGGGTCAG
- Phenotype
-
MIM: 614176
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MXD4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79484746] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MXD4
- Gene Name
- MAX dimerization protein 4
There are no transcripts associated with this gene.
- Gene
- ZFYVE28
- Gene Name
- zinc finger FYVE-type containing 28
View Full Product Details